missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-[2-(2,4-Difluorophenyl)-2,3-epoxypropyl]-1H-1,2,4-triazole methanesulfonate, 98%, Alfa Aesar™

Manufacturer:  Alfa Aesar H6178206

 View more versions of this product

Catalog No. AAH6178206



Specifications

Specifications

1-[2-(2,4-Difluorophenyl)-2,3-epoxypropyl]-1H-1,2,4-triazole methanesulfonate
86386-77-8
1-[2-(2,4-difluorophenyl)-2-oxiranylmethyl]-1H-1,2,4-triazole methanesulfonate
Solid
5g
MFCD11111978
333.31
98%

Chemical Identifiers

86386-77-8
MFCD11111978
1-[2-(2,4-difluorophenyl)-2-oxiranylmethyl]-1H-1,2,4-triazole methanesulfonate
Documents