Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-[2-(1-Pyrrolidinyl)ethyl]piperazine, 97%

Chemical Identifiers

Chemical Name or Material 1-[2-(1-Pyrrolidinyl)ethyl]piperazine
CAS 22763-69-5
Molecular Formula C10H21N3
Formula Weight 183.3
MDL Number MFCD01075190
UN Number UN2735
Synonym 1-[2-(1-Pyrrolidino)ethyl]piperazine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5069206

  SDS
alfa aesar™
H5069206
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 183.3
Boiling Point 77° to 79°C (0.5mmHg)
Quantity 5g
Molecular Formula C10H21N3
Health Hazard 3 GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
MDL Number MFCD01075190
Health Hazard 2 GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
UN Number UN2735
Synonym 1-[2-(1-Pyrrolidino)ethyl]piperazine
DOT Information Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.
Health Hazard 1 Danger
Chemical Name or Material 1-[2-(1-Pyrrolidinyl)ethyl]piperazine
CAS 22763-69-5
Assay 97%

Applications
1-[2-(1-Pyrrolidinyl)ethyl]piperazine is an important raw material and intermediate used in pharmaceuticals, agrochemicals and dyestuff.

Notes
Air Sensitive. Store away from air. Store under dry inert gas. Incompatible with air, carbondioxide and oxidizing agents.

Safety and Handling

Danger

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.