missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-(1H-Pyrrol-3-yl)ethan-1-one, 97%, Maybridge

Chemical Identifiers

CAS 1072-82-8
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Molecular Formula C6H7NO
Formula Weight 109.13
MDL Number MFCD00067759
Chemical Name or Material 1-(1H-pyrrol-3-yl)ethan-1-one
Assay 97%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
BTB07157EA SDS Fisher Scientific Company
BTB07157EA
Amber Glass Bottle 10g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

1072-82-8
100.0
C6H7NO
MFCD00067759
97%
96.5
97%
109.13
1-(1H-pyrrol-3-yl)ethan-1-one

Specifications

10g
96.5
Amber Glass Bottle
C6H7NO
MFCD00067759
97%
1072-82-8
100.0
97%
109.13
1-(1H-pyrrol-3-yl)ethan-1-one