missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

[1,1ft.-Bis(diphenylphosphino)ferrocene]palladium(II)chloride w/dichloromethane (1:1), Pd13%, AlfaAesar™

$72.04 - $72.04

Chemical Identifiers

CAS 95464-05-4
Physical Form Powder
Molecular Formula C34H28Cl2FeP2Pd·CH2Cl2
Formula Weight 816.64
MDL Number MFCD00792899
Synonym Dichloro[1, 1′-bis(diphenylphosphino)ferrocene]palladium(II) dichloromethane complex
Chemical Name or Material [1, 1′-Bis(diphenylphosphino)ferrocene]palladium(II) chloride
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA4122506 SDS Alfa Aesar
4122506
5g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AA4122514 SDS Alfa Aesar
4122514
25g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AA4122503 SDS Alfa Aesar
4122503
1g Each for $72.04
Specifications Description & Specifications

Chemical Identifiers

95464-05-4
C34H28Cl2FeP2Pd·CH2Cl2
MFCD00792899
[1, 1′-Bis(diphenylphosphino)ferrocene]palladium(II) chloride
Powder
816.64
Dichloro[1, 1′-bis(diphenylphosphino)ferrocene]palladium(II) dichloromethane complex

Specifications

5g
Powder
816.64
Dichloro[1, 1′-bis(diphenylphosphino)ferrocene]palladium(II) dichloromethane complex
95464-05-4
C34H28Cl2FeP2Pd·CH2Cl2
MFCD00792899
[1, 1′-Bis(diphenylphosphino)ferrocene]palladium(II) chloride