Please login to your online account to display your discounted pricing

1,14-Tetradecanediol 95%, ACROS Organics™


Quantity 500mg
Packaging Glass bottle

Manufacturer: thermo fisher scientific  420245000

 View more versions of this product

Catalog No. AC420245000

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥94%
Physical Form Crystals
Formula Weight 230.39
Quantity 500mg
Color White
EINECS Number 243-341-2
TSCA TSCA
Melting Point 85° to 89°C
CAS Min % 94.0
CAS Max % 100.0
Infrared Spectrum Authentic
Packaging Glass bottle
Health Hazard 1 HYGROSCOPIC
Chemical Name or Material 1, 14-Tetradecanediol
Name Note 95%
Molecular Formula C14H30O2
Linear Formula HOCH2(CH2)12CH2OH
MDL Number MFCD00004758
CAS 19812-64-7
Assay Percent Range 95%

Specifications

Chemical Name or Material 1, 14-Tetradecanediol
Name Note 95%
CAS 19812-64-7
CAS Min % 94.0
CAS Max % 100.0
Assay ≥94%
Assay Percent Range 95%
Molecular Formula C14H30O2
Linear Formula HOCH2(CH2)12CH2OH
Formula Weight 230.39
MDL Number MFCD00004758
Accessories

HYGROSCOPIC

SDS & Safety Documents