Please login to your online account to display your discounted pricing

1,14-Tetradecanediol 95%, ACROS Organics™


Quantity 100mg
Packaging Glass bottle

Manufacturer: thermo fisher scientific  420241000

 View more versions of this product

Catalog No. AC420241000

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Infrared Spectrum Authentic
Packaging Glass bottle
Color White
Melting Point 85° to 89°C
Quantity 100mg
Chemical Name or Material 1, 14-Tetradecanediol
Name Note 95%
CAS 19812-64-7
CAS Min % 94.0
CAS Max % 100.0
Physical Form White crystals
Assay 94% min.
Assay Percent Range 95%
Molecular Formula C14H30O2
Linear Formula HOCH2(CH2)12CH2OH
Formula Weight 230.39g/mol
MDL Number MFCD00004758
Health Hazard 1 HYGROSCOPIC
EINECS Number 243-341-2
TSCA TSCA

Specifications

Chemical Name or Material 1, 14-Tetradecanediol
Name Note 95%
CAS 19812-64-7
CAS Min % 94.0
CAS Max % 100.0
Assay 94% min.
Assay Percent Range 95%
Molecular Formula C14H30O2
Linear Formula HOCH2(CH2)12CH2OH
Formula Weight 230.39g/mol
MDL Number MFCD00004758
Accessories

HYGROSCOPIC

SDS & Safety Documents