Please login to your online account to display your discounted pricing

1,14-Tetradecanediol 95%, ACROS Organics™

$293.98 - $293.98

Chemical Identifiers

Chemical Name or Material 1, 14-Tetradecanediol
Name Note 95%
CAS 19812-64-7
CAS Min % 94.0
CAS Max % 100.0
Assay ≥94%
Assay Percent Range 95%
Molecular Formula C14H30O2
Linear Formula HOCH2(CH2)12CH2OH
Formula Weight 230.39
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC420241000

  SDS
thermo fisher scientific
420241000
Glass bottle 100mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC420245000

  SDS
thermo fisher scientific
420245000
Glass bottle 500mg Each for $293.98
Description & Specifications

Specifications

Assay ≥94%
Physical Form Crystals
Formula Weight 230.39
Quantity 100mg
Color White
EINECS Number 243-341-2
TSCA TSCA
Melting Point 85° to 89°C
CAS Min % 94.0
CAS Max % 100.0
Packaging Glass bottle
Infrared Spectrum Authentic
Health Hazard 1 HYGROSCOPIC
Chemical Name or Material 1, 14-Tetradecanediol
Molecular Formula C14H30O2
MDL Number MFCD00004758
CAS 19812-64-7
Name Note 95%
Assay Percent Range 95%
Linear Formula HOCH2(CH2)12CH2OH
Safety and Handling

HYGROSCOPIC