missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 1,13-Tetradecadiene, 90+%

Manufacturer:  Alfa Aesar™ H5347914

 View more versions of this product

Catalog No. AAH5347914



Specifications

Specifications

1,13-Tetradecadiene
131°C (17mmHg)
21964-49-8
CH2=CH(CH2)10CH=CH2
1741459
XMRSTLBCBDIKFI-UHFFFAOYSA-N
tetradeca-1,13-diene
30875
≥90%
103°C (217°F)
25g
C14H26
MFCD00014941
1,13-tetradecadiene
C=CCCCCCCCCCCC=C
194.362
194.36

Chemical Identifiers

21964-49-8
194.362
XMRSTLBCBDIKFI-UHFFFAOYSA-N
30875
C=CCCCCCCCCCCC=C
C14H26
MFCD00014941
1,13-tetradecadiene
tetradeca-1,13-diene
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

Documents