missing translation for 'onlineSavingsMsg'
Learn More

1,12-Dibromododecane, 98%

CAS: 3344-70-5 | C12H24Br2 | 328.132 g/mol

Supplier:  Thermo Scientific Chemicals B2125422

 View more versions of this product

Catalog No. AAB2125422


Only null left
Explore available promotions

Description

Description

1,12-Dibromododecane, is used as an organic chemical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,12-Dibromododecane, is used as an organic chemical intermediate.

Solubility
Soluble in hot methanol- almost transparency.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions.Stable. Combustible. Incompatible with strong oxidizing agents.
Specifications

Specifications

1,12-Dibromododecane
38°C to 42°C
>110°C (230°F)
MFCD00000226
1742763
Soluble in hot methanol- almost transparency.
C(CCCCCCBr)CCCCCBr
328.132
328.13
3344-70-5
215°C (15 mmHg)
C12H24Br2
100 g
dodecamethylene dibromide, dodecane, 1,12-dibromo, alpha,omega-dibromododecane, labotest-bb ltbb002671, pubchem3879, 1, 12-dibromododecane, 1,12-dibromo-dodecane, 1,12-bis bromanyl dodecane, 4-01-00-00503 beilstein handbook reference, ksc225m2h
ZJJATABWMGVVRZ-UHFFFAOYSA-N
1,12-dibromododecane
18766
98%

Chemical Identifiers

3344-70-5
328.132
ZJJATABWMGVVRZ-UHFFFAOYSA-N
18766
C(CCCCCCBr)CCCCCBr
C12H24Br2
MFCD00000226
dodecamethylene dibromide, dodecane, 1,12-dibromo, alpha,omega-dibromododecane, labotest-bb ltbb002671, pubchem3879, 1, 12-dibromododecane, 1,12-dibromo-dodecane, 1,12-bis bromanyl dodecane, 4-01-00-00503 beilstein handbook reference, ksc225m2h
1,12-dibromododecane
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : 222-096-5

RTECSNumber : JR2295000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only