missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
1,12-Dibromododecane, 96%, ACROS Organics™
Manufacturer: Acros Organics AC158455000
Specifications
1,12-Dibromododecane | |
Authentic | |
215.0°C (15.0 mmHg) | |
37.0°C to 42.0°C | |
3344-70-5 | |
100.0 | |
C12H24Br2 | |
MFCD00000226 | |
dodecamethylene dibromide, dodecane, 1,12-dibromo, alpha,omega-dibromododecane, labotest-bb ltbb002671, pubchem3879, 1, 12-dibromododecane, 1,12-dibromo-dodecane, 1,12-bis bromanyl dodecane, 4-01-00-00503 beilstein handbook reference, ksc225m2h | |
C(CCCCCCBr)CCCCCBr | |
328.132 | |
328.13 | |
96% |
>110°C | |
Glass bottle | |
White to Brown | |
500g | |
95.0 | |
96% | |
Br(CH2)12Br | |
01, II, 134 | |
ZJJATABWMGVVRZ-UHFFFAOYSA-N | |
1,12-dibromododecane | |
18766 | |
Crystalline Solid |
Chemical Identifiers
3344-70-5 | |
328.132 | |
ZJJATABWMGVVRZ-UHFFFAOYSA-N | |
18766 | |
C(CCCCCCBr)CCCCCBr |
C12H24Br2 | |
MFCD00000226 | |
dodecamethylene dibromide, dodecane, 1,12-dibromo, alpha,omega-dibromododecane, labotest-bb ltbb002671, pubchem3879, 1, 12-dibromododecane, 1,12-dibromo-dodecane, 1,12-bis bromanyl dodecane, 4-01-00-00503 beilstein handbook reference, ksc225m2h | |
1,12-dibromododecane |