missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1,12-Dibromododecane, 96%, ACROS Organics™

Manufacturer:  Acros Organics AC158450050

 View more versions of this product

Catalog No. AC158450050


 

Specifications

Specifications

1,12-Dibromododecane
215.0°C (15.0 mmHg)
37.0°C to 42.0°C
3344-70-5
95.0
Glass bottle
96%
Br(CH2)12Br
01, II, 134
ZJJATABWMGVVRZ-UHFFFAOYSA-N
1,12-dibromododecane
18766
Crystalline Solid
>110°C
White through Brown
5g
Authentic
100.0
96%
C12H24Br2
MFCD00000226
Dodecamethylene dibromide
C(CCCCCCBr)CCCCCBr
328.132
328.13

Chemical Identifiers

3344-70-5
328.132
ZJJATABWMGVVRZ-UHFFFAOYSA-N
18766
C12H24Br2
MFCD00000226
Dodecamethylene dibromide
SDS Documents