missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
1,11-Dodecadiene 99.0+%, TCI America™
$349.57 - $1273.87
Chemical Identifiers
CAS | 5876-87-9 |
---|---|
Molecular Formula | C12H22 |
Molecular Weight (g/mol) | 166.308 |
MDL Number | MFCD00048156 |
InChI Key | IYPLTVKTLDQUGG-UHFFFAOYSA-N |
PubChem CID | 22170 |
IUPAC Name | dodeca-1,11-diene |
SMILES | C=CCCCCCCCCC=C |
Chemical Identifiers
5876-87-9 | |
166.308 | |
IYPLTVKTLDQUGG-UHFFFAOYSA-N | |
dodeca-1,11-diene |
C12H22 | |
MFCD00048156 | |
22170 | |
C=CCCCCCCCCC=C |
Specifications
5876-87-9 | |
207°C | |
MFCD00048156 | |
IYPLTVKTLDQUGG-UHFFFAOYSA-N | |
dodeca-1,11-diene | |
22170 | |
≥99.0% (GC) | |
1,11-Dodecadiene |
Yellow | |
C12H22 | |
5 mL | |
C=CCCCCCCCCC=C | |
166.308 | |
166.31 | |
Liquid |
Safety and Handling
TSCA : No