missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

(1,10-Phenanthroline)(trifluoromethyl)(triphenylphosphine)copper(I), TCI America™

Manufacturer:  TCI America P21725G

 View more versions of this product

Catalog No. P21725G



Specifications

Specifications

(1,10-Phenanthroline)(trifluoromethyl)(triphenylphosphine)copper(I)
1334890-93-5
C31H23CuF3N2P
C1=CC=C(C=C1)P(C2=CC=CC=C2)C3=CC=CC=C3.C1=CC2=C(C3=C(C=CC=N3)C=C2)N=C1.[C-](F)(F)F.[Cu+]
575.054
575.05
Undesignated
5g
≥95.0% (T)
AKGOGTMOALVICI-UHFFFAOYSA-N
copper(1+);1,10-phenanthroline;trifluoromethane;triphenylphosphane
88921701
Crystal-Powder at 20°C

Chemical Identifiers

1334890-93-5
575.054
88921701
C1=CC=C(C=C1)P(C2=CC=CC=C2)C3=CC=CC=C3.C1=CC2=C(C3=C(C=CC=N3)C=C2)N=C1.[C-](F)(F)F.[Cu+]
C31H23CuF3N2P
AKGOGTMOALVICI-UHFFFAOYSA-N
copper(1+);1,10-phenanthroline;trifluoromethane;triphenylphosphane
Safety and Handling

Safety and Handling

TSCA : No

Documents