Please login to your online account to display your discounted pricing

1,10-Phenanthroline Monohydrate (Certified ACS), Fisher Chemical

$172.62 - $218.38

Chemical Identifiers

Chemical Name or Material 1, 10-Phenanthroline Monohydrate
CAS 5144-89-8
Purity Grade Certified ACS Grade
Molecular Formula C12H8N2 · H2O
Formula Weight 198.23
MDL Number MFCD00149973
Synonym o-Phenanthroline
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

P70-10

  SDS
fisher scientific company
P7010
Glass Bottle 10g Each for $218.38
Case of 6 Each for $1,310.28

P70-5

  SDS
fisher scientific company
P705
Glass Bottle 5g Each for $172.62
Case of 6 Each for $1,035.72
Description & Specifications

Specifications

Purity Grade Certified ACS Grade
Physical Form Powder/Solid
Formula Weight 198.23
Melting Point 97°C
Quantity 10g
DOT Information DOT Class 6.1, : Poison
Molecular Formula C12H8N2 · H2O
Viscosity [NULL]
Alpha Vector Phenanthroline Monohydrate
Color Off-white
Packaging Glass Bottle
MDL Number MFCD00149973
ChemAlert Storage Symbol Blue
Suitability Pass Test (as redox indicator and for Fe test)
CAS 5144-89-8
Synonym o-Phenanthroline
Chemical Name or Material 1, 10-Phenanthroline Monohydrate
Safety and Handling