missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1,10-Phenanthroline, Monohydrate, ACS Grade, LabChem™

$133.51 - $133.51

Chemical Identifiers

CAS 5144-89-8
CAS Max % 100
Linear Formula C12H8N2·H2O
Formula Weight 198.22
Chemical Name or Material 1,10-Phenanthroline, Monohydrate
Grade ACS Grade
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
LC181407 Lab Chem
LC181407
Amber Glass 5g Each for $133.51
Specifications Description & Specifications

Chemical Identifiers

5144-89-8
C12H8N2·H2O
1,10-Phenanthroline, Monohydrate
100
198.22
ACS Grade

Specifications

5g
100
Amber Glass
C12H8N2·H2O
Soluble in water
Room Temperature
Carbon dioxide; Carbon monoxide; Nitrogen oxides
White
1,10-Phenanthroline, Monohydrate
5144-89-8
Passes Test
Crystalline Powder
198.22
1100kg/m3
Passes Test
1,100kg/m3
93°C
ACS Grade

Safety and Handling

GHS H Statement
Toxic if swallowed.
Very toxic to aquatic life with long lasting effects.

GHS P Statement
Wash exposed skin thoroughly after handling.
Do not eat, drink or smoke when using this product.
If swallowed: Rinse mouth.
Immediately call a poison center/doctor.
Avoid release to the environment.
Collect spillage.
Dispose of contents/container to comply with local, state and federal regulations.

Danger

EINECSNumber : 200-629-2