Learn More
1,10-Phenanthroline monohydrate, ACS, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 03091018
Description
1,10-Phenanthroline monohydrate is useful to study the DNA-protein interactions due to its nuclease activity. It is used as a redox indicator and reagent for spectrophotometric assay of silver. It acts as a chelating ligand for the determination of Fe, Pd and V. It also acts as a matrix metalloproteinase inhibitor. It is a photometric reagent for iron, which forms a red chelate with ferrous (iron II) ion.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
1,10-Phenanthroline monohydrate | |
97°C to 101°C | |
C12H10N2O | |
50 g | |
4008096 | |
1,10-phenanthroline hydrate, 1,10-phenanthroline monohydrate, o-phenanthroline monohydrate, unii-ksx215x00e, 1,10-fenantrolina, 4,5-phenanthroline monohydrate, 1,10-phenanthroline, monohydrate, 1,10-fenanthrolin, 1,10-phenanthrolineo-phenanthroline, 1, 10-phenanthroline monohydrate | |
PPQJCISYYXZCAE-UHFFFAOYSA-N | |
1,10-phenanthroline;hydrate | |
21226 | |
ACS |
5144-89-8 | |
Odorless | |
MFCD00149973 | |
UN2811 | |
14,7214 | |
Soluble in alcohol,acetone; Slightly soluble in waterSoluble in ethanol,acetone and methanol; Slightly soluble in water and benzene. | |
O.C1=CN=C2C(C=CC3=CC=CN=C23)=C1 | |
198.23 | |
198.23 (180.21 Anhydrous) | |
Crystalline |
Chemical Identifiers
5144-89-8 | |
198.23 | |
PPQJCISYYXZCAE-UHFFFAOYSA-N | |
21226 | |
O.C1=CN=C2C(C=CC3=CC=CN=C23)=C1 |
C12H10N2O | |
MFCD00149973 | |
1,10-phenanthroline hydrate, 1,10-phenanthroline monohydrate, o-phenanthroline monohydrate, unii-ksx215x00e, 1,10-fenantrolina, 4,5-phenanthroline monohydrate, 1,10-phenanthroline, monohydrate, 1,10-fenanthrolin, 1,10-phenanthrolineo-phenanthroline, 1, 10-phenanthroline monohydrate | |
1,10-phenanthroline;hydrate |
Safety and Handling
GHS H Statement
H301
Toxic if swallowed.
P264b-P270-P301+P310-P330-P501c
H301
DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
EINECSNumber : 200-629-2
RTECSNumber : SF8437000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only