missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 1,10-Phenanthroline iron(II) sulfate, 0.025M aq. soln.

Manufacturer:  Alfa Aesar™ 3056218

 View more versions of this product

Catalog No. AA3056218



Specifications

Specifications

1,10-Phenanthroline iron(II) sulfate
50g
C36H24FeN6O4S
ferroin, ferroin indicator solution, iron 2+ , tris 1,10-phenanthroline-.kappa.n1,.kappa.n10-, oc-6-11-, sulfate 1:1, ferroin sulfate, ferroin solution, 1,10-phenanthroline ferrous sulfate, tris 1,10-phenanthroline-n1,n10 iron sulphate, ferrous 1,10-phenanthroline sulfate, ferroin indicator solution, 25 mmol/l, iron 2+ , tris 1,10-phenanthroline-kappan1,kappan10-, oc-6-11-, sulfate 1:1
CIWXFRVOSDNDJZ-UHFFFAOYSA-L
iron(2+);1,10-phenanthroline;sulfate
84567
Liquid
Red
14634-91-4
MFCD00036428
0.025M aq. soln.
C1=CC2=C(C3=C(C=CC=N3)C=C2)N=C1.C1=CC2=C(C3=C(C=CC=N3)C=C2)N=C1.C1=CC2=C(C3=C(C=CC=N3)C=C2)N=C1.[O-]S(=O)(=O)[O-].[Fe+2]
692.531
692.53

Chemical Identifiers

14634-91-4
692.531
CIWXFRVOSDNDJZ-UHFFFAOYSA-L
84567
C1=CC2=C(C3=C(C=CC=N3)C=C2)N=C1.C1=CC2=C(C3=C(C=CC=N3)C=C2)N=C1.C1=CC2=C(C3=C(C=CC=N3)C=C2)N=C1.[O-]S(=O)(=O)[O-].[Fe+2]
C36H24FeN6O4S
MFCD00036428
ferroin, ferroin indicator solution, iron 2+ , tris 1,10-phenanthroline-.kappa.n1,.kappa.n10-, oc-6-11-, sulfate 1:1, ferroin sulfate, ferroin solution, 1,10-phenanthroline ferrous sulfate, tris 1,10-phenanthroline-n1,n10 iron sulphate, ferrous 1,10-phenanthroline sulfate, ferroin indicator solution, 25 mmol/l, iron 2+ , tris 1,10-phenanthroline-kappan1,kappan10-, oc-6-11-, sulfate 1:1
iron(2+);1,10-phenanthroline;sulfate
Safety and Handling

Safety and Handling

TSCA : TSCA

SDS Documents