missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 1,10-Phenanthroline iron(II) perchlorate

Oxidation, reduction indicator

Chemical Identifiers

CAS 14586-54-0
Formula Weight 795.37
MDL Number MFCD00050959
UN Number UN1479
Chemical Name or Material 1,10-Phenanthroline iron(II) perchlorate
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA3055709 SDS Alfa Aesar™
3055709
10g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

14586-54-0
MFCD00050959
1,10-Phenanthroline iron(II) perchlorate
795.37
UN1479

Specifications

10g
Crystalline
MFCD00050959
Red
14586-54-0
795.37
UN1479
1,10-Phenanthroline iron(II) perchlorate