Learn More
1,10-Phenanthroline, 99+%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 157530250
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
1, 10-Phenanthroline | |
99.0 | |
117.0°C to 120.0°C | |
Authentic | |
Glass bottle | |
MFCD00011678 | |
23,227 | |
o-phenanthroline, 4,5-diazaphenanthrene, orthophenanthroline, phenanthroline, 1,10-o-phenanthroline, phen, 2-phenanthroline, beta-phenanthroline, anhydrous, .beta.-phenanthroline | |
DGEZNRSVGBDHLK-UHFFFAOYSA-N | |
1,10-phenanthroline | |
1318 | |
180.21 | |
Powder |
66-71-7 | |
100.0 | |
Pink or White to Yellow | |
99+% | |
C12H8N2 | |
25 g | |
15,7328 | |
Solubility in water: slightly soluble. Other solubilities: soluble in alcohol, acetone, ether and benzene | |
C1=CC2=C(C3=C(C=CC=N3)C=C2)N=C1 | |
180.21 | |
CHEBI:44975 | |
99+% |
Chemical Identifiers
66-71-7 | |
180.21 | |
DGEZNRSVGBDHLK-UHFFFAOYSA-N | |
1318 | |
1,10-phenanthroline |
C12H8N2 | |
MFCD00011678 | |
o-phenanthroline, 4,5-diazaphenanthrene, orthophenanthroline, phenanthroline, 1,10-o-phenanthroline, phen, 2-phenanthroline, beta-phenanthroline, anhydrous, .beta.-phenanthroline | |
CHEBI:44975 | |
C1=CC2=C(C3=C(C=CC=N3)C=C2)N=C1 |
Safety and Handling
GHS H Statement:
Very toxic to aquatic life with long lasting effects.
Toxic if swallowed.
GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Avoid release to the environment.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
EINECSNumber : 200-629-2
RTECSNumber : SF8300000
TSCA : TSCA
RUO – Research Use Only