Learn More
1,10-Phenanthroline-5,6-dione, 98%, Thermo Scientific Chemicals
$117.70 - $1324.40
Chemical Identifiers
CAS | 27318-90-7 |
---|---|
Molecular Formula | C12H6N2O2 |
Molecular Weight (g/mol) | 210.19 |
MDL Number | MFCD00014473 |
InChI Key | KCALAFIVPCAXJI-UHFFFAOYSA-N |
Synonym | 1,10 phenanthroline-5,6-dione, 1 10-phenanthroline-5 6-dione 97, pyridino 3,2-h quinoline-5,6-dione, phen-5,6-dione, pubchem16755, acmc-209gwd, d0jh7c, phox31, ksc204s8l, 1,2-dione-based compound, 17 |
PubChem CID | 72810 |
IUPAC Name | 1,10-phenanthroline-5,6-dione |
SMILES | O=C1C(=O)C2=CC=CN=C2C2=NC=CC=C12 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAH55747MD
|
Thermo Scientific Chemicals
H55747MD |
250 mg |
Each for $117.70
|
|
|||||
AAH5574703
|
Thermo Scientific Chemicals
H5574703 |
1 g |
Each for $310.25
|
|
|||||
AAH5574706
|
Thermo Scientific Chemicals
H5574706 |
5 g |
Each for $1,324.40
|
|
|||||
Description
1,10-Phenanthroline-5,6-dione is a Bifunctional quinone oxidant which, when used in conjunction with Zn2+ catalysts, is used to affect the aerobic oxidation of secondary amines to a variety of value added motifs, including indoles.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications1,10-Phenanthroline-5,6-dione is a Bifunctional quinone oxidant which, when used in conjunction with Zn2+ catalysts, is used to affect the aerobic oxidation of secondary amines to a variety of value added motifs, including indoles.
Solubility
Insoluble in water.
Notes
Store in a cool and dark place. Store away from incompatible materials such as oxidizing agents.
Chemical Identifiers
27318-90-7 | |
210.19 | |
KCALAFIVPCAXJI-UHFFFAOYSA-N | |
72810 | |
O=C1C(=O)C2=CC=CN=C2C2=NC=CC=C12 |
C12H6N2O2 | |
MFCD00014473 | |
1,10 phenanthroline-5,6-dione, 1 10-phenanthroline-5 6-dione 97, pyridino 3,2-h quinoline-5,6-dione, phen-5,6-dione, pubchem16755, acmc-209gwd, d0jh7c, phox31, ksc204s8l, 1,2-dione-based compound, 17 | |
1,10-phenanthroline-5,6-dione |
Specifications
27318-90-7 | |
C12H6N2O2 | |
250 mg | |
Insoluble in water. | |
O=C1C(=O)C2=CC=CN=C2C2=NC=CC=C12 | |
210.19 | |
210.19 | |
1,10-Phenanthroline-5,6-dione |
260°C (decomposition) | |
MFCD00014473 | |
1,10 phenanthroline-5,6-dione, 1 10-phenanthroline-5 6-dione 97, pyridino 3,2-h quinoline-5,6-dione, phen-5,6-dione, pubchem16755, acmc-209gwd, d0jh7c, phox31, ksc204s8l, 1,2-dione-based compound, 17 | |
KCALAFIVPCAXJI-UHFFFAOYSA-N | |
1,10-phenanthroline-5,6-dione | |
72810 | |
98% |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only