missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 1,10-Phenanthroline-5,6-dione, 98%

Manufacturer:  Alfa Aesar™ H55747MD

 View more versions of this product

Catalog No. AAH55747MD



Description

Description

Applications
1,10-Phenanthroline-5,6-dione is a Bifunctional quinone oxidant which, when used in conjunction with Zn2+ catalysts, is used to affect the aerobic oxidation of secondary amines to a variety of value added motifs, including indoles.

Solubility
Insoluble in water.

Notes
Store in a cool and dark place. Store away from incompatible materials such as oxidizing agents.
Specifications

Specifications

1,10-Phenanthroline-5,6-dione
250mg
C12H6N2O2
1 10-phenanthroline-5 6-dione 97
KCALAFIVPCAXJI-UHFFFAOYSA-N
1,10-phenanthroline-5,6-dione
72810
98%
260°C (decomposition)
27318-90-7
MFCD00014473
Insoluble in water.
C1=CC2=C(C3=C(C=CC=N3)C(=O)C2=O)N=C1
210.192
210.19

Chemical Identifiers

27318-90-7
210.192
KCALAFIVPCAXJI-UHFFFAOYSA-N
72810
C1=CC2=C(C3=C(C=CC=N3)C(=O)C2=O)N=C1
C12H6N2O2
MFCD00014473
1 10-phenanthroline-5 6-dione 97
1,10-phenanthroline-5,6-dione
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

Documents