Please login to your online account to display your discounted pricing

1,10-Phenanthroline, 0.3% (w/v) Aqueous Solution, Ricca Chemical


Quantity 100mL
Packaging Poly natural

Manufacturer: ricca chemical company  RIR5522000100A

 View more versions of this product

Catalog No. R5522000100

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Chemical Name or Material 1, 10-Phenanthroline
CAS 7732-18-5
CAS Min % 98.0
CAS Max % 100.0
CAS 5144-89-8
CAS Min % 0.29
CAS Max % 0.3
CAS 7647-01-0
CAS Min % 0.12
CAS Max % 0.12
Physical Form Liquid
Packaging Poly natural
Concentration or Composition Notes 0.3% (w/v)
Solvent or Matrix Aqueous Solution
Color Clear colorless to very light green
Quantity 100mL

Specifications

Chemical Name or Material 1, 10-Phenanthroline
CAS 7732-18-5
CAS Min % 98.0
CAS Max % 100.0
CAS 5144-89-8
CAS Min % 0.29
CAS Max % 0.3
CAS 7647-01-0
CAS Min % 0.12
CAS Max % 0.12
Physical Form Liquid