missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
1,10-Dibromodecane, 97%, ACROS Organics™
Manufacturer: Thermo Fisher Scientific AC112785000
Specifications
1, 10-Dibromodecane | |
>112°C | |
Glass bottle | |
1.34 | |
Beige to Brown | |
500mL | |
96.0 | |
96% min. (GC) | |
Br(CH2)10Br | |
01, I, 64 | |
Solubility in water: insoluble | |
C(CCCCCBr)CCCCBr | |
300.07 | |
300.07 | |
97% |
1.3400g/mL | |
Authentic | |
1.4912 | |
160°C (15.0mmHg) | |
25°C to 30°C | |
4101-68-2 | |
100.0 | |
C10H20Br2 | |
MFCD00000222 | |
1,10 dibromodecane, 1,10-decamethylene bromide, 1,10-decamethylene dibromide, 1,10-dibromdecane, 1,10-dibromo-decan, 1,10-dibromodecan, a,w-dibromodecane, decamethylene dibromide, decane, 1,10-dibromo, decane,10-dibromo | |
GTQHJCOHNAFHRE-UHFFFAOYSA-N | |
1,10-dibromodecane | |
221483 | |
Crystalline Low Melting Solid |
Chemical Identifiers
4101-68-2 | |
300.07 | |
GTQHJCOHNAFHRE-UHFFFAOYSA-N | |
221483 | |
C(CCCCCBr)CCCCBr |
C10H20Br2 | |
MFCD00000222 | |
1,10 dibromodecane, 1,10-decamethylene bromide, 1,10-decamethylene dibromide, 1,10-dibromdecane, 1,10-dibromo-decan, 1,10-dibromodecan, a,w-dibromodecane, decamethylene dibromide, decane, 1,10-dibromo, decane,10-dibromo | |
1,10-dibromodecane |
Safety and Handling
EINECSNumber : 223-871-
Recommended Storage : Product may darken during storage