Please login to your online account to display your discounted pricing

Alfa Aesar™ 1,10-Dibromodecane, 97%

$498.74 - $498.74

Chemical Identifiers

Chemical Name or Material 1,10-Dibromodecane
CAS 4101-68-2
Molecular Formula Br(CH2)10Br
Formula Weight 300.08
MDL Number MFCD00000222
Beilstein 506156
Synonym Decamethylene dibromide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1235736

  SDS
alfa aesar™
A1235736
500g Each for $498.74
Description & Specifications

Specifications

Formula Weight 300.08
CAS 4101-68-2
Synonym Decamethylene dibromide
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Boiling Point 157° to 159°C (14mmHg)
MDL Number MFCD00000222
Health Hazard 1 Warning
Molecular Formula Br(CH2)10Br
Melting Point 26° to 30°C
Beilstein 506156
TSCA TSCA
Quantity 500g
Flash Point >110°C (230°F)
Chemical Name or Material 1,10-Dibromodecane
Assay 97%

Applications
1,10-Dibromodecane is used as an alkylating agent and as synthetic organic intermediate.

Solubility
Insoluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with bases and oxidizing agents.

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.