Please login to your online account to display your discounted pricing

1,10-Diamino-4,7-dioxadecane, 95%, ACROS Organics™

$100.99 - $100.99

Chemical Identifiers

Chemical Name or Material 1, 10-Diamino-4, 7-dioxadecane
CAS 2997-01-5
CAS Min % 94.0
CAS Max % 100.0
Assay ≥94%
Assay Percent Range 95%
Molecular Formula C8H20N2O2
Linear Formula NH2(CH2)3O(CH2)2O(CH2)3NH2
Formula Weight 176.26
MDL Number MFCD00059938
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC371610250

  SDS
thermo fisher scientific
AC371610250
Glass bottle 25g Each for $100.99

AC371610050

  SDS
thermo fisher scientific
AC371610050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥94%
Physical Form Liquid
Formula Weight 176.26
Quantity 25g
Color Clear colorless
Density 0.9837g/mL
CAS Min % 94.0
CAS Max % 100.0
Linear Formula NH2(CH2)3O(CH2)2O(CH2)3NH2
CAS 2997-01-5
MDL Number MFCD00059938
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 2 GHS H Statement:
Harmful in contact with skin.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Assay Percent Range 95%
Infrared Spectrum Authentic
Packaging Glass bottle
Refractive Index 1.4620 to 1.4640 (20°C, 589nm)
Chemical Name or Material 1, 10-Diamino-4, 7-dioxadecane
Health Hazard 1 GHS Signal Word: Danger
Molecular Formula C8H20N2O2
Safety and Handling

GHS Signal Word: Danger

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful in contact with skin.
Causes severe skin burns and eye damage.
Causes serious eye damage.