missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
1,10-Decanediol, 99%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 181045000
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
1, 10-Decanediol | |
98.5 | |
297°C | |
Authentic | |
Glass bottle | |
HO(CH2)10OH | |
500g | |
15, 2853 | |
Solubility in water: insoluble in water. Other solubilities: freely soluble in alcohol and warm ether,practically insoluble in petroleum ether | |
C(CCCCCO)CCCCO | |
174.28 | |
174.28 |
112-47-0 | |
100.0 | |
152°C | |
98.5% min. (GC) | |
C10H22O2 | |
MFCD00004749 | |
01, II, 560 | |
1,10-decanediol, decamethylene glycol, decamethylenediol, 1,10-dihydroxydecane, 1,10-decamethylenediol, 1,10-decamethylene diol, 1,6-bis 2-hydroxyethyl hexane, alpha,omega-decanediol, unii-5i577udk52, 1,10-ddecanediol | |
FOTKYAAJKYLFFN-UHFFFAOYSA-N | |
decane-1,10-diol | |
37153 | |
99% |
Safety and Handling
EINECSNumber : 203-975-2
RUO â Research Use Only