Please login to your online account to display your discounted pricing

Alfa Aesar™ 1,10-Decanedicarbonitrile, 98%

$97.74 - $420.23

Chemical Identifiers

Chemical Name or Material 1,10-Decanedicarbonitrile
CAS 4543-66-2
Molecular Formula NC(CH2)10CN
Formula Weight 192.3
MDL Number MFCD00013818
Beilstein 1757752
Synonym 1,10-Dicyanodecane; Dodecanedinitrile
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0345106

 
alfa aesar™
L0345106
5g Each for $97.74

AAL0345114

 
alfa aesar™
L0345114
25g Each for $420.23
Description & Specifications

Specifications

Formula Weight 192.3
Chemical Name or Material 1,10-Decanedicarbonitrile
CAS 4543-66-2
Assay 98%
Molecular Formula NC(CH2)10CN
MDL Number MFCD00013818
Beilstein 1757752
Quantity 5g
Synonym 1,10-Dicyanodecane; Dodecanedinitrile
RTECS Number JR2350000
Boiling Point 195°C (8mmHg)
Melting Point 16° to 19°C
Flash Point 202°C (395°F)

Applications
The Dodecanedinitrile could react with C-tert-butoxy-tetra-N-methyl-methanediamine to obtain the 2-(Dimethylaminomethylen)dodecansaeuredinitril.

Solubility
Insoluble in water.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.