Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-(1-Propynyl)cyclohexanol, 98%

$61.26 - $197.03

Chemical Identifiers

Chemical Name or Material 1-(1-Propynyl)cyclohexanol
CAS 697-37-0
Molecular Formula C9H14O
Formula Weight 138.21
MDL Number MFCD00078311
Beilstein 1856828
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2203809

 
alfa aesar™
B2203809
10g Each for $61.26

AAB2203818

 
alfa aesar™
B2203818
50g Each for $197.03
Description & Specifications

Specifications

Formula Weight 138.21
Chemical Name or Material 1-(1-Propynyl)cyclohexanol
Assay 98%
Molecular Formula C9H14O
MDL Number MFCD00078311
CAS 697-37-0
Beilstein 1856828
Boiling Point 91° to 92°C (10mmHg)
Melting Point 46° to 50°C
Quantity 10g