Please login to your online account to display your discounted pricing

Alfa Aesar™ 1,1'-Oxybis(3,3,3-trifluoro-1-propanol) hemihydrate, 96%

Chemical Identifiers

Chemical Name or Material 1,1'-Oxybis(3,3,3-trifluoro-1-propanol) hemihydrate
CAS 936107-87-8
Molecular Formula C6H8F6O3·0.5H2O
Linear Formula [CF3CH2CH(OH)]2O·0.5H2O
Formula Weight 251.12 (242.12 Anhydrous)
MDL Number MFCD00156074
UN Number UN1988
Synonym 3,3,3-Trifluoropropionaldehyde hemihydrate
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5169003

  SDS
alfa aesar™
H5169003
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH5169006

  SDS
alfa aesar™
H5169006
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 251.12 (242.12 Anhydrous)
Chemical Name or Material 1,1'-Oxybis(3,3,3-trifluoro-1-propanol) hemihydrate
Assay 96%
Molecular Formula C6H8F6O3·0.5H2O
Linear Formula [CF3CH2CH(OH)]2O·0.5H2O
MDL Number MFCD00156074
CAS 936107-87-8
UN Number UN1988
Synonym 3,3,3-Trifluoropropionaldehyde hemihydrate
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H331-H318-H226-H302-H315-H335
Toxic if inhaled.
Causes serious eye damage.
Flammable liquid and vapor.
Harmful if swallowed.
Causes skin irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ALDEHYDES, FLAMMABLE, TOXIC, N.O.S.
Quantity 1g
Safety and Handling

Danger

GHS P Statement
P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H331-H318-H226-H302-H315-H335
Toxic if inhaled.
Causes serious eye damage.
Flammable liquid and vapor.
Harmful if swallowed.
Causes skin irritation.
May cause respiratory irritation.