missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1,1′-Oxalyldiimidazole 95.0+%, TCI America™

$81.31 - $254.86

Chemical Identifiers

CAS 18637-83-7
Molecular Formula C8H6N4O2
Molecular Weight (g/mol) 190.162
MDL Number MFCD00043067
InChI Key ONRNRVLJHFFBJG-UHFFFAOYSA-N
Synonym 1,1'-oxalyl-diimidazol, 1,1'-oxalyldi 1h-imidazole, 1,1'-oxalyldiimidazole, 1,1-oxalyldiimidazole, 1,2-di 1h-imidazol-1-yl ethane-1,2-dione, 1,2-di imidazol-1-yl ethane-1,2-dione, 1h-imidazole, 1,1'-1,2-dioxo-1,2-ethanediyl bis, acmc-1cacg, bis imidazol-1-yl ethane-1,2-dione, oxalic acid diimidazolide
PubChem CID 100439
IUPAC Name 1,2-di(imidazol-1-yl)ethane-1,2-dione
SMILES C1=CN(C=N1)C(=O)C(=O)N2C=CN=C2
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
O02001G
View Documents
Tci America
O02001G
1g
Each for $81.31
 
O02005G
View Documents
Tci America
O02005G
5g
Each for $254.86
 
Description

Description

Specifications

Chemical Identifiers

18637-83-7
190.162
ONRNRVLJHFFBJG-UHFFFAOYSA-N
100439
C1=CN(C=N1)C(=O)C(=O)N2C=CN=C2
C8H6N4O2
MFCD00043067
1,1'-oxalyl-diimidazol, 1,1'-oxalyldi 1h-imidazole, 1,1'-oxalyldiimidazole, 1,1-oxalyldiimidazole, 1,2-di 1h-imidazol-1-yl ethane-1,2-dione, 1,2-di imidazol-1-yl ethane-1,2-dione, 1h-imidazole, 1,1'-1,2-dioxo-1,2-ethanediyl bis, acmc-1cacg, bis imidazol-1-yl ethane-1,2-dione, oxalic acid diimidazolide
1,2-di(imidazol-1-yl)ethane-1,2-dione

Specifications

Undesignated
18637-83-7
MFCD00043067
ONRNRVLJHFFBJG-UHFFFAOYSA-N
1,2-di(imidazol-1-yl)ethane-1,2-dione
100439
Crystal-Powder at 20°C
1,1′-Oxalyldiimidazole
1g
C8H6N4O2
1,1'-oxalyl-diimidazol, 1,1'-oxalyldi 1h-imidazole, 1,1'-oxalyldiimidazole, 1,1-oxalyldiimidazole, 1,2-di 1h-imidazol-1-yl ethane-1,2-dione, 1,2-di imidazol-1-yl ethane-1,2-dione, 1h-imidazole, 1,1'-1,2-dioxo-1,2-ethanediyl bis, acmc-1cacg, bis imidazol-1-yl ethane-1,2-dione, oxalic acid diimidazolide
C1=CN(C=N1)C(=O)C(=O)N2C=CN=C2
190.162
190.16
≥95.0% (T)
Safety and Handling

Safety and Handling

TSCA : No

SDS