missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
1-(1-Naphthyl)ethylamine, 98%, ACROS Organics™
Manufacturer: Thermo Fisher Scientific AC450571000
Specifications
1-(1-Naphthyl)ethylamine | |
>110°C | |
1.063 | |
100g | |
98% | |
+/--1-1-naphthyl ethylamine, +/-1-1-naphthyl ethylamine, 1-1-naphthyl ethanamine, 1-1-naphthyl ethylamine, 1-1-napthyl ethylamine, 1-naphthalen-1-yl ethan-1-amine, 1-naphthalen-1-yl ethanamine, 1-naphthalen-1-yl-ethylamine, dl-1-1-naphthyl ethylamine, r-+-1-1-naphthyl ethylamine | |
RTCUCQWIICFPOD-UHFFFAOYSA-N | |
1-naphthalen-1-ylethanamine | |
98089 | |
98% |
1.0630g/mL | |
Glass bottle | |
156°C (15.0mmHg) | |
42882-31-5 | |
C12H13N | |
Solubility in water: insoluble | |
CC(C1=CC=CC2=CC=CC=C21)N | |
171.24 | |
171.24 |
Chemical Identifiers
42882-31-5 | |
171.24 | |
+/--1-1-naphthyl ethylamine, +/-1-1-naphthyl ethylamine, 1-1-naphthyl ethanamine, 1-1-naphthyl ethylamine, 1-1-napthyl ethylamine, 1-naphthalen-1-yl ethan-1-amine, 1-naphthalen-1-yl ethanamine, 1-naphthalen-1-yl-ethylamine, dl-1-1-naphthyl ethylamine, r-+-1-1-naphthyl ethylamine | |
1-naphthalen-1-ylethanamine |
C12H13N | |
RTCUCQWIICFPOD-UHFFFAOYSA-N | |
98089 | |
CC(C1=CC=CC2=CC=CC=C21)N |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
GHS Signal Word: Warning