Please login to your online account to display your discounted pricing

1-(1-Naphthyl)ethylamine, 98%, Acros Organics


Quantity 25g

Manufacturer: thermo fisher scientific  AC450570250

 View more versions of this product

Catalog No. AC450570250

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Chemical Name or Material 1-(1-Naphthyl)ethylamine
CAS 42882-31-5
Packaging Glass bottle
Assay Percent Range 98%
Molecular Formula C12H13N
Formula Weight 171.24
Quantity 25g

Specifications

Chemical Name or Material 1-(1-Naphthyl)ethylamine
Assay Percent Range 98%
Molecular Formula C12H13N
Formula Weight 171.24