missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
1-(1-Naphthyl)-2-(trimethylsilyl)acetylene, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals H5595314
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
1-(1-Naphthyl)-2-(trimethylsilyl)acetylene | |
41°C to 43°C | |
292°C to 293°C | |
C15H16Si | |
MFCD04039886 | |
1-1-naphthyl-2-trimethylsilyl acetylene, trimethyl 2-naphthalen-1-ylethynyl silane, trimethyl 2-naphthalen-1-yl ethynyl silane, 1-naphthylethynyl trimethylsilane, 1-trimethylsilylethynyl naphthalene, 1-trimethylsilyl ethynyl naphthalene, trimethyl naphthalen-1-ylethynyl silane, trimethyl naphthalen-1-yl ethynyl silane, trimethylnaphthalen-1-ylethynylsilane | |
C[Si](C)(C)C#CC1=CC=CC2=CC=CC=C21 | |
224.378 | |
224.38 | |
Solid |
104784-51-2 | |
0.968 g/mL | |
>110°C (230°F) | |
1.595 | |
25 g | |
WATBCTJRCLNXNF-UHFFFAOYSA-N | |
trimethyl(2-naphthalen-1-ylethynyl)silane | |
4438232 | |
97% |
Chemical Identifiers
104784-51-2 | |
224.378 | |
WATBCTJRCLNXNF-UHFFFAOYSA-N | |
4438232 | |
C[Si](C)(C)C#CC1=CC=CC2=CC=CC=C21 |
C15H16Si | |
MFCD04039886 | |
1-1-naphthyl-2-trimethylsilyl acetylene, trimethyl 2-naphthalen-1-ylethynyl silane, trimethyl 2-naphthalen-1-yl ethynyl silane, 1-naphthylethynyl trimethylsilane, 1-trimethylsilylethynyl naphthalene, 1-trimethylsilyl ethynyl naphthalene, trimethyl naphthalen-1-ylethynyl silane, trimethyl naphthalen-1-yl ethynyl silane, trimethylnaphthalen-1-ylethynylsilane | |
trimethyl(2-naphthalen-1-ylethynyl)silane |
Safety and Handling
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only