Please login to your online account to display your discounted pricing

Alfa Aesar™ 1,1'-Ferrocenedicarboxylic acid, 98%

$56.58 - $189.19

Chemical Identifiers

Chemical Name or Material 1,1'-Ferrocenedicarboxylic acid
CAS 1293-87-4
Molecular Formula C12H10FeO4
Formula Weight 274.06g/mol
MDL Number MFCD00001423
Synonym 1,1'-Dicarboxyferrocene; Bis(carboxycyclopentadienyl)iron
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1669103

 
alfa aesar™
A16691-03
1g Each for $56.58

AAA1669106

 
alfa aesar™
A16691-06
5g Each for $189.19
Description & Specifications

Specifications

Melting Point >300°C
Quantity 1g
Odor Odorless
Chemical Name or Material 1,1'-Ferrocenedicarboxylic acid
CAS 1293-87-4
Assay 98%
Molecular Formula C12H10FeO4
Formula Weight 274.06g/mol
MDL Number MFCD00001423
Synonym 1,1'-Dicarboxyferrocene; Bis(carboxycyclopentadienyl)iron
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.