Please login to your online account to display your discounted pricing

Alfa Aesar™ 1,1'-Ferrocenedicarboxaldehyde, 97%

$206.27 - $652.65

Chemical Identifiers

Chemical Name or Material 1,1'-Ferrocenedicarboxaldehyde
CAS 1271-48-3
Molecular Formula C12H10FeO2
Formula Weight 242.06
MDL Number MFCD02093629
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA4429202

 
alfa aesar™
4429202
0.5g Each for $206.27

AA4429204

 
alfa aesar™
4429204
2g Each for $652.65
Description & Specifications

Specifications

Formula Weight 242.06
Chemical Name or Material 1,1'-Ferrocenedicarboxaldehyde
Physical Form Powder
Assay 97%
Molecular Formula C12H10FeO2
MDL Number MFCD02093629
CAS 1271-48-3
Sensitivity Air Sensitive
Melting Point 183° to 184°C
Quantity 0.5g