missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 1,1'-Ferrocenedicarboxaldehyde, 97%

Manufacturer:  Alfa Aesar™ 4429204

 View more versions of this product

Catalog No. AA4429204



Specifications

Specifications

1,1'-Ferrocenedicarboxaldehyde
183°C to 184°C
1271-48-3
MFCD02093629
242.06
97%
Air sensitive
2g
C12H10FeO2
Insoluble in water.
Powder

Chemical Identifiers

1271-48-3
C12H10FeO2
MFCD02093629
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Ambient temperatures

Documents