Please login to your online account to display your discounted pricing

Alfa Aesar™ 1,1'-Ethynylenedicyclohexanol, 97%

$363.27 - $363.27

Chemical Identifiers

Chemical Name or Material 1,1'-Ethynylenedicyclohexanol
CAS 78-54-6
Molecular Formula C14H22O2
Formula Weight 222.33
MDL Number MFCD00021400
Synonym Bis(1-hydroxycyclohexyl)acetylene
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1948822

  SDS
alfa aesar™
A1948822
100g Each for $363.27
Description & Specifications

Specifications

Formula Weight 222.33
Chemical Name or Material 1,1'-Ethynylenedicyclohexanol
Assay 97%
Molecular Formula C14H22O2
MDL Number MFCD00021400
CAS 78-54-6
Synonym Bis(1-hydroxycyclohexyl)acetylene
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Quantity 100g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.