Please login to your online account to display your discounted pricing

Alfa Aesar™ 1,1-Diphenylpentane, 97%

$206.27 - $206.27

Chemical Identifiers

Chemical Name or Material 1,1-Diphenylpentane
CAS 1726-12-1
Molecular Formula (C6H5)2CH(CH2)3CH3
Formula Weight 224.35
MDL Number MFCD00041692
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2355814

 
alfa aesar™
B2355814
25g Each for $206.27
Description & Specifications

Specifications

Formula Weight 224.35
Chemical Name or Material 1,1-Diphenylpentane
Assay 97%
Molecular Formula (C6H5)2CH(CH2)3CH3
MDL Number MFCD00041692
CAS 1726-12-1
Boiling Point 138° to 139°C (15mmHg)
Quantity 25g