Please login to your online account to display your discounted pricing

Alfa Aesar™ 1,1-Diphenylhexane, 98%

$328.47 - $328.47

Chemical Identifiers

Chemical Name or Material 1,1-Diphenylhexane
CAS 1530-04-7
Molecular Formula (C6H5)2CH(CH2)4CH3
Formula Weight 238.38
MDL Number MFCD00041694
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2289614

  SDS
alfa aesar™
B2289614
25g Each for $328.47

AAB2289606

  SDS
alfa aesar™
B2289606
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material 1,1-Diphenylhexane
CAS 1530-04-7
Assay 98%
Molecular Formula (C6H5)2CH(CH2)4CH3
Formula Weight 238.38
MDL Number MFCD00041694
Boiling Point 129° to 130°C (2mmHg)
Quantity 25g