Please login to your online account to display your discounted pricing

Alfa Aesar™ 1,1-Dioxobenzo[b]thiophen-2-ylmethyl N-succimidyl carbonate, 95%

$62.96 - $243.20

Chemical Identifiers

Chemical Name or Material 1,1-Dioxobenzo[b]thiophen-2-ylmethyl N-succimidyl carbonate
CAS 197244-91-0
Molecular Formula C14H11NO7S
Formula Weight 337.31
MDL Number MFCD03095506
Beilstein 8274967
Synonym N-(Benzo[b]thiophenesulfone-2-methoxycarbonyl)succinimide; Bsmoc-OSu
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1973303

  SDS
alfa aesar™
L1973303
1g Each for $62.96

AAL1973306

  SDS
alfa aesar™
L1973306
5g Each for $243.20
Description & Specifications

Specifications

Formula Weight 337.31
Chemical Name or Material 1,1-Dioxobenzo[b]thiophen-2-ylmethyl N-succimidyl carbonate
Molecular Formula C14H11NO7S
CAS 197244-91-0
Synonym N-(Benzo[b]thiophenesulfone-2-methoxycarbonyl)succinimide; Bsmoc-OSu
Beilstein 8274967
Melting Point 140°C
Quantity 1g
Assay 95%
MDL Number MFCD03095506
Recommended Storage 2° to 8°C
Safety and Handling

2° to 8°C