Please login to your online account to display your discounted pricing

1,1-Dimethylurea +98%, ACROS Organics™


Quantity 100g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC388741000

 View more versions of this product

Catalog No. AC388741000

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥98.0%
Physical Form Crystalline Powder
Formula Weight 88.11
Quantity 100g
Color White to off-white
Melting Point 179° to 183°C
CAS Min % 98.0
CAS Max % 100.0
Molecular Formula C3H8N2O
Linear Formula (CH3)2NC(O)nH2
Assay Percent Range 98+%
CAS 598-94-7
MDL Number MFCD00007959
Packaging Glass bottle
Chemical Name or Material 1, 1-Dimethylurea
Infrared Spectrum Authentic

Specifications

Chemical Name or Material 1, 1-Dimethylurea
CAS 598-94-7
CAS Min % 98.0
CAS Max % 100.0
Assay ≥98.0%
Assay Percent Range 98+%
Molecular Formula C3H8N2O
Linear Formula (CH3)2NC(O)nH2
Formula Weight 88.11
MDL Number MFCD00007959