Please login to your online account to display your discounted pricing

Alfa Aesar™ 1,1'-Dimethylferrocene, 97%

$24.14 - $269.97

Chemical Identifiers

Chemical Name or Material 1,1'-Dimethylferrocene
CAS 1291-47-0
Molecular Formula C12H14Fe
Formula Weight 214.09g/mol
MDL Number MFCD00001425
Synonym Bis(methylcyclopentadienyl)iron
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1217503

 
alfa aesar™
A12175-03
1g Each for $24.14

AAA1217514

 
alfa aesar™
A12175-14
25g Each for $269.97
Description & Specifications

Specifications

Melting Point 36° to 41°C
Quantity 1g
Flash Point 108°C (226°F)
Chemical Name or Material 1,1'-Dimethylferrocene
CAS 1291-47-0
Assay 97%
Molecular Formula C12H14Fe
Formula Weight 214.09g/mol
MDL Number MFCD00001425
Synonym Bis(methylcyclopentadienyl)iron

Applications
It is used as a combustion control additive in fuels, antiknock agent in gasoline, and for heat stabilization in greases and plastics. It is used as a catalyst for synthesis of ammonia, polymerization.

Solubility
Slightly soluble in methanol. Insoluble in water.

Notes
Store in cool, dry place in a tightly sealed container. Store away from oxidizing agent.