Please login to your online account to display your discounted pricing

Alfa Aesar™ (1,1-Dimethoxyethyl)benzene, 97%

Chemical Identifiers

Chemical Name or Material (1,1-Dimethoxyethyl)benzene
CAS 4316-35-2
Formula Weight 166.22
MDL Number MFCD00075617
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5569606

  SDS
alfa aesar™
H5569606
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH5569614

  SDS
alfa aesar™
H5569614
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Boiling Point 95° to 98°C (12mmHg)
Formula Weight 166.22
Quantity 5g
Density 1.008g/mL
TSCA TSCA
Refractive Index 1.4900
MDL Number MFCD00075617
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H227
Combustible liquid.
Color Colorless
CAS 4316-35-2
Health Hazard 3 GHS P Statement
P210-P280-P370+P378a-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
In case of fire: Use for extinction: CO2, powder or water spray.
Store in a well-ventilated place.
Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Physical Form Liquid
Assay 97%
Chemical Name or Material (1,1-Dimethoxyethyl)benzene
Safety and Handling

Warning

GHS P Statement
P210-P280-P370+P378a-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
In case of fire: Use for extinction: CO2, powder or water spray.
Store in a well-ventilated place.
Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H227
Combustible liquid.