missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 1,1'-Diethyl-4,4'-carbocyanine iodide, 96%

Manufacturer:  Alfa Aesar™ H5545603

 View more versions of this product

Catalog No. AAH5545603



Specifications

Specifications

1,1'-Diethyl-4,4'-carbocyanine iodide
Green
1g
C25H25IN2
UN2811
CEJANLKHJMMNQB-UHFFFAOYSA-M
(4E)-1-ethyl-4-[(Z)-3-(1-ethylquinolin-1-ium-4-yl)prop-2-enylidene]quinoline;iodide
16219292
480.38
96%
Light sensitive
250°C to 253°C
4727-50-8
MFCD00011970
1,1'-diethylquino-4-carbocyanine iodide
CCN1C=CC(=CC=CC2=CC=[N+](C3=CC=CC=C23)CC)C4=CC=CC=C41.[I-]
480.393
CHEBI:51502
Powder

Chemical Identifiers

4727-50-8
480.393
CEJANLKHJMMNQB-UHFFFAOYSA-M
16219292
(4E)-1-ethyl-4-[(Z)-3-(1-ethylquinolin-1-ium-4-yl)prop-2-enylidene]quinoline;iodide
C25H25IN2
MFCD00011970
1,1'-diethylquino-4-carbocyanine iodide
CHEBI:51502
CCN1C=CC(=CC=CC2=CC=[N+](C3=CC=CC=C23)CC)C4=CC=CC=C41.[I-]
Safety and Handling

Safety and Handling

GHS H Statement
H300-H315-H319-H335
Fatal if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P311-P312-P330-P332+P313-P361-P363-P501c

H300-H311+H331-H315-H319

DOTInformation : Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.

EINECSNumber : 225-224-8

RTECSNumber : VC3676880

TSCA : Yes

Recommended Storage : Ambient temperatures

Documents