Learn More
1,1'-Diethyl-4,4'-carbocyanine iodide, 96%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals H55456MD
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
1,1'-Diethyl-4,4'-carbocyanine iodide | |
250°C to 253°C | |
C25H25IN2 | |
250 mg | |
Light sensitive | |
CEJANLKHJMMNQB-UHFFFAOYSA-M | |
(4E)-1-ethyl-4-[(Z)-3-(1-ethylquinolin-1-ium-4-yl)prop-2-enylidene]quinoline;iodide | |
16219292 | |
480.38 | |
Powder |
4727-50-8 | |
Green | |
MFCD00011970 | |
UN2811 | |
cryptocyanin, cryptocyanine, kryptocyanin, kryptocyanine, cryptocyanine iodide, kryptocyanine iodide, 1,1'-diethylquino-4-carbocyanine iodide, cryptocyanine o.a. 2, 1,4'-carbocyanine iodide, 1,4'-quinocarbocyanine iodide | |
CCN1C=CC(=CC=CC2=CC=[N+](C3=CC=CC=C23)CC)C4=CC=CC=C41.[I-] | |
480.393 | |
CHEBI:51502 | |
96% |
Chemical Identifiers
4727-50-8 | |
480.393 | |
CEJANLKHJMMNQB-UHFFFAOYSA-M | |
16219292 | |
(4E)-1-ethyl-4-[(Z)-3-(1-ethylquinolin-1-ium-4-yl)prop-2-enylidene]quinoline;iodide |
C25H25IN2 | |
MFCD00011970 | |
cryptocyanin, cryptocyanine, kryptocyanin, kryptocyanine, cryptocyanine iodide, kryptocyanine iodide, 1,1'-diethylquino-4-carbocyanine iodide, cryptocyanine o.a. 2, 1,4'-carbocyanine iodide, 1,4'-quinocarbocyanine iodide | |
CHEBI:51502 | |
CCN1C=CC(=CC=CC2=CC=[N+](C3=CC=CC=C23)CC)C4=CC=CC=C41.[I-] |
Safety and Handling
GHS H Statement
H300-H315-H319-H335
Fatal if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P311-P312-P330-P332+P313-P361-P363-P501c
H300-H311+H331-H315-H319
DOTInformation : Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
EINECSNumber : 225-224-8
RTECSNumber : VC3676880
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only