missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 1,1'-Diethyl-2,2'-cyanine iodide, 97%

Manufacturer:  Alfa Aesar™ H55022MD

 View more versions of this product

Catalog No. AAH55022MD



Description

Description

Applications
1,1′-Diethyl-2,2′-cyanine iodide is used to inhibit 1-methyl-4-phenylpyridinium (MPP+), toxic metabolite of 1-methyl-4-phenyl-1,2,3,6-tetrahydropyridine that induces apoptosis in cerebellar granule neurons (CGNs).

Solubility
Soluble to 10 mM in dimethyl sulfoxide.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents. It is sensitive to light.
Specifications

Specifications

1,1'-Diethyl-2,2'-cyanine iodide
273°C (decomposition)
977-96-8
MFCD00011971
decynium 22
GMYRVMSXMHEDTL-UHFFFAOYSA-M
(2Z)-1-ethyl-2-[(1-ethylquinolin-1-ium-2-yl)methylidene]quinoline;iodide
71299759
454.35
Light sensitive
250mg
C23H23IN2
UN2811
Soluble to 10 mM in dimethyl sulfoxide.
CCN1C(=CC2=[N+](C3=CC=CC=C3C=C2)CC)C=CC4=CC=CC=C41.[I-]
454.355
CHEBI:37993
97%

Chemical Identifiers

977-96-8
454.355
GMYRVMSXMHEDTL-UHFFFAOYSA-M
71299759
(2Z)-1-ethyl-2-[(1-ethylquinolin-1-ium-2-yl)methylidene]quinoline;iodide
C23H23IN2
MFCD00011971
decynium 22
CHEBI:37993
CCN1C(=CC2=[N+](C3=CC=CC=C3C=C2)CC)C=CC4=CC=CC=C41.[I-]
Safety and Handling

Safety and Handling

GHS H Statement
H300-H311-H330-H315-H319-H335
Fatal if swallowed.
Toxic in contact with skin.
Fatal if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P260-P264b-P270-P271-P280-P284-P301+P310-P302+P352-P304+P340-P305+P351+P338-P310-P312-P330-P332+P313-P362-P501c

H300+H330-H312-H315-H319-H335

DOTInformation : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.

EINECSNumber : 213-556-6

TSCA : Yes

Recommended Storage : Ambient temperatures

Documents