Please login to your online account to display your discounted pricing

Alfa Aesar™ 1,1-Dichloro-2-phenylcyclopropane, 97%

$238.07 - $789.54

Chemical Identifiers

Chemical Name or Material 1,1-Dichloro-2-phenylcyclopropane
CAS 2415-80-7
Molecular Formula C9H8Cl2
Formula Weight 187.07
MDL Number MFCD00001272
Beilstein 1860351
Synonym (2,2-Dichlorocyclopropyl)benzene
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1227914

  SDS
alfa aesar™
A1227914
25g Each for $238.07

AAA1227922

  SDS
alfa aesar™
A1227922
100g Each for $789.54
Description & Specifications

Specifications

Formula Weight 187.07
CAS 2415-80-7
Flash Point 104°C (219°F)
Synonym (2,2-Dichlorocyclopropyl)benzene
Beilstein 1860351
Boiling Point 105° to 106°C (12mmHg)
Chemical Name or Material 1,1-Dichloro-2-phenylcyclopropane
Molecular Formula C9H8Cl2
MDL Number MFCD00001272
Assay 97%
Quantity 25g