Please login to your online account to display your discounted pricing

Alfa Aesar™ 1,1-Dibromopinacolone, 98%

$69.27 - $1,042.09

Chemical Identifiers

Chemical Name or Material 1,1-Dibromopinacolone
CAS 30263-65-1
Molecular Formula C6H10Br2O
Linear Formula (CH3)3CCOCHBr2
Formula Weight 257.95
MDL Number MFCD00045030
Synonym 1,1-Dibromo-3,3-dimethylbutan-2-one
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1651718

 
alfa aesar™
A1651718
50g Each for $252.45

AAA1651709

 
alfa aesar™
A1651709
10g Each for $69.27

AAA1651730

 
alfa aesar™
A1651730
250g Each for $1,042.09
Description & Specifications

Specifications

Formula Weight 257.95
Chemical Name or Material 1,1-Dibromopinacolone
Assay 98%
Molecular Formula C6H10Br2O
Linear Formula (CH3)3CCOCHBr2
MDL Number MFCD00045030
CAS 30263-65-1
Synonym 1,1-Dibromo-3,3-dimethylbutan-2-one
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Recommended Storage 2° to 8°C
Boiling Point 96° to 97°C (12mmHg)
Quantity 50g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

2° to 8°C