Please login to your online account to display your discounted pricing

Alfa Aesar™ 1,1'-Dibromoferrocene, 96%

$53.88 - $152.38

Chemical Identifiers

Chemical Name or Material 1,1'-Dibromoferrocene
CAS 1293-65-8
Molecular Formula C10H8Br2Fe
Formula Weight 343.83g/mol
MDL Number MFCD01075755
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1704503

 
alfa aesar™
L17045-03
1g Each for $152.38

AAL17045MD

 
alfa aesar™
L17045-MD
250mg Each for $53.88
Description & Specifications

Specifications

Melting Point 50° to 51°C
Quantity 1g
Chemical Name or Material 1,1'-Dibromoferrocene
CAS 1293-65-8
Assay 96%
Molecular Formula C10H8Br2Fe
Formula Weight 343.83g/mol
MDL Number MFCD01075755
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Recommended Storage 2° to 8°C
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

2° to 8°C