missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1,1-Dibromoethane, 99%, stabilized, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 406481000

 View more versions of this product

Catalog No. AC406481000

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

≥98.5%
187.86
Clear colorless to brown
100.0
98.5
Glass bottle
Ethylidene bromide
C2H4Br2
99%
1, 1-Dibromoethane
Liquid
100g
2.0600g/mL
potassium carbonate
1.5090 to 1.5120 (20°C, 589nm)
Authentic
99%
557-91-5
CH3CHBr2
MFCD00041719

Chemical Identifiers

557-91-5
100.0
C2H4Br2
187.86
Ethylidene bromide
≥98.5%
98.5
99%
CH3CHBr2
MFCD00041719
1, 1-Dibromoethane, 99%

Safety and Handling

SDS & Safety Documents

GHS H Statement:
Harmful in contact with skin.
Suspected of causing cancer.
Harmful if inhaled.
Harmful if swallowed.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
Use personal protective equipment as required.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning

EINECSNumber : 209-184-9

RTECSNumber : KH9000000

TSCA : TSCA